ContactUsLinkComponent
Tempo iPeri™  Human iPSC derived Pericytes
Tempo iPeri™  Human iPSC derived Pericytes
# de catalogue 76682-968
Fournisseur:  TEMPO BIOSCIENCE, INC
Numéro CAS:  
IPERIHUMAN IPSC-DERIVED PERICYTES
Tempo iPeri™  Human iPSC derived Pericytes
# de catalogue 76682-968
Fournisseur:  TEMPO BIOSCIENCE, INC
Numéro CAS:  
Spécifications
  • Description:
    Human iPSC-derived Pericytes
Spécifications
A propos de cet article
Human iPSC-derived Pericytes.
  • Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought