Tempo iMG™ Human iPSC derived Microglia
# de catalogue 76682-934
Fournisseur: TEMPO BIOSCIENCE, INC
Numéro CAS:
Spécifications
- Description:Human iPSC-derived microglia
Spécifications
A propos de cet article
Human iPSC-derived Microglia.
- Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought