TempoDual™ ATPMito Rapid Dual Biosensor Cytotoxicity Assay
# de catalogue 76682-956
Fournisseur: TEMPO BIOSCIENCE, INC
Numéro CAS:
Spécifications
- Description:Dual TempoATP-TempoMito biosensor cytotoxicity assay
Spécifications
A propos de cet article
Dual TempoATP-TempoMito Biosensor Cytotoxicity Assay.
- Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought