TempoVol™-iCort: Cationic Voltage Biosensor Assay using Human Cortical Neurons
# de catalogue 76682-990
Fournisseur: TEMPO BIOSCIENCE, INC
Numéro CAS:
Spécifications
- Description:Cationic voltage biosensor assay using human cortical Neurons
Spécifications
A propos de cet article
Cationic Voltage Biosensor Assay using Human Cortical Neurons.
- Cryovial. ~1 million cells/ml
Product Type: human cells
Long-Term Storage: liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought