TempoMito™ iAstro Mitochondrial Biosensor Assay using Human Astrocytes
# de catalogue 76682-942
Fournisseur: TEMPO BIOSCIENCE, INC
Numéro CAS:
Spécifications
- Description:Mitochondrial biosensor assay using human astrocytes
Spécifications
A propos de cet article
Mitochondrial Biosensor Assay using Human Astrocytes.
- Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought