Tempo-iKer™: Human iPSC-derived Keratinocytes
# de catalogue 76683-020
Fournisseur: TEMPO BIOSCIENCE, INC
Numéro CAS:
Spécifications
- Description:Human iPSC-derived Keratinocytes
Spécifications
A propos de cet article
Human iPSC-derived Keratinocytes.
- Cryovial. ~1 million cells/ml
Product Type: human cells
Long-Term Storage: liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought