TempoMito™ Rapid Mitochondrial Biosensor Assay using Immortalized Human Neuroepithelial Cells
# de catalogue 76682-912
Fournisseur: TEMPO BIOSCIENCE, INC
Numéro CAS:
Spécifications
- Description:TempoMito biosensor transiently expressed in TempoRapid neuroepithelial cells
Spécifications
A propos de cet article
TempoMito biosensor transiently expressed in TempoRapid neuroepithelial cells
- 1 Cryovial = ~1.025×10^⁶
- cells/ml
Product Type: human cells
Long-Term Storage: liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2
Customers Who Bought This Also Bought