ContactUsLinkComponent
AimStrip® Xylazine Urine/Powder Dipstick, Germaine Laboratories
AimStrip® Xylazine Urine/Powder Dipstick, Germaine Laboratories
# de catalogue 76717-408
Fournisseur:  Germaine Laboratories
XYLAZINE DIPSTICK AIMSTRIP PK25
AimStrip® Xylazine Urine/Powder Dipstick, Germaine Laboratories
# de catalogue 76717-408
Fournisseur:  Germaine Laboratories
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    AimStrip™ Xylazine Urine Dipstick
  • Size:
    25/Kit
  • Cat. no.:
    76717-408
Spécifications
A propos de cet article
AimStrip® Xylazine Urine/Powder Dipstick, 25/Kit. Cut-off concentration of 1000 ng/ml.
  • XYL has been reported to be misused as a horse doping agent, a drug of abuse and as source of accidental or intended poisonings.
  • Human intoxication with XYL has been reported sporadically over the past several decades
  • Noted in literature describing drug overdose deaths in Philadelphia as early as 2006
  • Since mid-2010's, XYL has been noticed by people who inject drugs (PWID) and public health practitioners as an increasingly commonplace additive in the street opioid supply of Philadelphia.
  • Recent reports from Connecticut implicated XYL in a rising fraction of overdose deaths in 2019–2020
  • In Puerto Rico, heroin is commonly adulterated with XYL
  • Frequently found in speedball (a cocaine and heroin mixture)
A rapid test for the qualitative detection of xylazine in human urine and powder. For forensic use only, not for in vitro diagnostic use. A rapid chromatographic immunoassay for the detection of xylazine in human urine and powders at a cut-off concentration of 1000 ng/ml. The Xylazine (XYL) Rapid Test Dipstick is a rapid screening test that can be performed without the use of an instrument. The test utilizes the antibody to selectively detect elevated levels of xylazine in urine and powders.
XYL has been reported to be misused as a horse doping agent, a drug of abuse and as source of accidental or intended poisonings.
Delivery: Ships ground, can ship expedited if specified on PO or requested timely with [email protected]
Caution: For forensic use only, not for in vitro diagnostic use
Customers Who Bought This Also Bought