Eisco Premium Qualitative Filter Papers
Fournisseur: Wards
Certificats
A propos de cet article
Pack of 100 premium Qualitative Filter Papers with medium speed (85 gsm) and 10 µm pore size.
- High retentivity, perfect for general laboratory filtering and qualitative analysis
- 10 µm (micron) pore size
- Medium speed (85 gsm)
Premium quality filter papers for use in labs. Pack of 100 circles.
Customers Who Bought This Also Bought