Corepoint® Scientific Hazardous Location Laboratory Refrigerators
Supplier: Horizon Scientific
Certificates
About this item
These refrigerators are designed to keep you and your stored substances safe in work areas where hazardous conditions may exist.
- Laboratory use
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These Hazardous Location Laboratory Refrigerators meet NFPA and OSHA guidelines for Hazardous Location, Class 1, Division II, Groups C and D guidelines. These units utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist.
Models feature a microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.
Certifications: ETL listed
*Select models ENERGY STAR certified: 76717-356
*Select models ENERGY STAR certified: 76717-356
Specifications
- Catalog No:
- 76717-356
- 76717-354
- 76717-358
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76717-356
- 76717-354
- 76717-358
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Shelves/drawers:
- 5 Adjustable shelves (4 full/1 half shelf)
- 5 Adjustable shelves (4 full/1 half shelf)
- 5 Adjustable shelves (4 full/1 half shelf)
- Door type:
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- Weight:
- 102 kg (225 lbs.)
- 82 kg (180 lbs.)
- 107 kg (235 lbs.)