Corepoint® Scientific White Diamond Flammable Material Storage Touchscreen Manual Defrost Freezers
Supplier: Horizon Scientific
Certificates
About this item
Safeguard volatile substances and upgrade your performance with these specially-designed flammable storage freezers.
- Laboratory use
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These White Diamond Series Flammable Storage Freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers.
This White Diamond Line showcases features including state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL listed
*Select models ENERGY STAR certified: 76717-360, 76717-364, and 76717-366
*Select models ENERGY STAR certified: 76717-360, 76717-364, and 76717-366
Ordering information: Three-year parts and labor warranty plus an additional two-year compressor parts warranty.
Specifications
- Catalog No:
- 76717-360
- 76717-366
- 76717-364
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76717-360
- 76717-366
- 76717-364
- Defrost:
- Manual
- Manual
- Manual
- Shelves/drawers:
- 7 Inner doors
- 7 Inner doors
- 7 Inner doors
- Door type:
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- Weight:
- 91 kg (200 lbs.)
- 119 kg (262 lbs.)
- 113 kg (250 lbs.)