Corepoint® Scientific General Purpose Laboratory Refrigerators
Supplier: Horizon Scientific
Certificates
About this item
These General Purpose Refrigerators are ideal for laboratory applications.
- Laboratory use
- Microprocessor temperature controller
- Spacious shelving
- Multiple size options
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
General Purpose Laboratory Refrigerators take a large step forward in performance from past general purpose units; offering a digital temperature display and alarm module coupled with a microprocessor temperature controller.
Certifications: ETL listed
*Select models ENERGY STAR certified: 76717-390
*Select models ENERGY STAR certified: 76717-390
Specifications
- Catalog No:
- 76717-392
- 76717-390
- 76717-388
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76717-392
- 76717-390
- 76717-388
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Shelves/drawers:
- 7 Adjustable shelves
- 6 Adjustable shelves
- 5 Adjustable shelves
- Door type:
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- Weight:
- 107 kg (235 lbs.)
- 102 kg (225 lbs.)
- 82 kg (180 lbs.)