Density Cube Set of 24
# de catalogue 470303-938
Fournisseur: United Scientific Supplies
Spécifications
- Description:Density cube set of 24
- Cat. no.:470303-938
Spécifications
A propos de cet article
Observe the density across different materials and sizes.
- Foam storage space
- Includes a Vernier caliper
This set includes three different sized cubes of eight materials commonly used in density experiments. Each material is represented by a 0.5, 1 and 1.5" cube. Materials included are acrylic, aluminum, brass, copper, nylon, polypropylene, PVC and steel. Set includes a reference table showing density values of individual specimens.
Customers Who Bought This Also Bought