Thermo Scientific™ Piercing Plate for Matrix™ 2D Barcoded Storage Tubes, 384
# de catalogue 76642-936
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Piercing plate, Matrix 384, 2D tubes
- Cat. no.:76642-936
Spécifications
A propos de cet article
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.
Customers Who Bought This Also Bought