Hand-Held Weather Center
# de catalogue 470332-742
Fournisseur: Learning Resources
Spécifications
- Description:Weather center box
- Cat. no.:470332-742
Spécifications
A propos de cet article
Easily carry around this economical weather station!
This set of three weather-measuring instruments includes its own carrying case and includes a 3' barometer (for measuring atmospheric pressure), hygrometer (for measuring humidity), and a Celsius and Fahrenheit thermometer. Individual stands are also included in the case. A helpful guide explains how to use each instrument.
Customers Who Bought This Also Bought