A propos de cet article
Maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily with the Thermo Scientific™ Capit-All™ screw cap tube capper/ decapper.
- Increase throughput by capping and decapping an entire rack at a time in approximately 10 seconds
- Automatically remove and replace caps on tubes of differing heights, even in the same rack, as well as tubes not properly seated in racks
- Completely replace manual capping and decapping methods
Designed for benchtop or integrated use, Capit-All instruments cap and decap 24-, 48-, and 96- format Thermo Scientific™ Matrix™ and Thermo Scientific™ Nunc™ Screw top tubes with precision and ease. All instruments cap tubes to optimal torque.
Superior sample protection:
Caps all tubes in a rack to the optimal torque level, maintaining sample integrity and preventing sample loss during storage
Safety features ensure racks are loaded in the correct orientation and that caps are not cross-threaded or overtightened
Drip tray prevents cross contamination of samples during addition and removal of tube racks
Internal construction ensures debris is not introduced into uncapped tubes
Ease of use:
The simple, four-button user interface enables laboratory personnel of any level to operate the instrument
Easily integrated into automated systems
Easily maintained and able to process up to a million tubes before requiring preventative maintenance