Thermo Scientific™ 8-Channel Screw Cap Decapper
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Process a full rack of Thermo Scientific™ Matrix™ or Thermo Scientific™ Nunc™ tubes in less than one minute with this ergonomic, manual capper/decapper.
- The Thermo Scientific™ 8-Channel Screw Cap Capper/Decapper is designed to cap tubes to the optimal torque to maintain sample integrity and prevent sample loss during storage
The 8-channel Capper/Decapper is a practical addition to benchtop processing of screw-top storage tubes.
Customers Who Bought This Also Bought