Adjustable Funnel Stand
# de catalogue 470343-566
Fournisseur: Wards
Spécifications
- Description:Adjustable funnel stand
- Cat. no.:470343-566
Spécifications
A propos de cet article
Crafted from polished wood, this stand with a 1.5" hole diameter and a height of 17" provides stability and convenience for various funnel applications.
- Funnel stand featuring a sturdyhardwood construction that will stand up to the wear and tear of the lab
- Height extends up to 18"
- Base is approximately 7³/₄" by 4¹/₄"
- Stand comes with clamping screws for adjusting the height
- Designed for use in professional laboratories, but is an ideal choice for science classrooms
Funnel stand made from quality polished wood. Hole diameter is 1.5". Height of the funnels is easily adjustable. Features a clamping screw to ensure easy movement when loose and sturdy placement when tightened, and a sturdy wooden base with rubber stoppers for increased stability. Each funnel stand is 12" long, 4.25" wide, and stands 17" tall. Minimum height of funnel attachment is 3" above the base, maximum height is 16" above the base.
Customers Who Bought This Also Bought