Spécifications
- Description:Polypropylene graduated cylinders set
- Cat. no.:470345-330
Spécifications
A propos de cet article
Set includes eight polypropylene graduated cylinders with hexagonal base, raised graduations, and varying capacities (10 to 2000 ml), offering precise measurement and stability for lab work.
- Polypropylene graduated cylinders
- Hexagonal base
- Autoclavable
- Perfect classroom or lab starter set
- Labeled graduations
Polypropylene graduated cylinders, set of 8, hexagonal base, labeled Graduations - 10, 25, 50, 100, 250, 500, 1000, 2000 ml eight cylinders These polypropylene graduated cylinders have a hexagonal base, are made from a single piece and are autoclavable. This is a perfect cylinder starter set including the following: 10 ml capacity, 0.2 ml graduation 25 ml capacity, 0.5 ml graduation 50 ml capacity, 1.0 ml graduation 100 ml capacity, 1.0 ml graduation 250 ml capacity, 2.0 ml graduation 500 ml capacity, 5.0 ml graduation 1000 ml capacity, 10.0 ml graduation 2000 ml capacity, 20.0 ml graduation calibrated to contain.