9-Piece Organic Chemistry Kit
# de catalogue 470343-812
Fournisseur: Wards
Spécifications
- Description:9-piece organic chemistry kit
- Cat. no.:470343-812
Spécifications
A propos de cet article
With a 14/23 joint size, this organic chemistry kit consists of 9 essential pieces made of borosilicate glass, providing the necessary tools for a range of experiments and ensuring durability and chemical resistance.
- This set contains 9 items with 14/23 ground joints for maximum versatility
- Glassware pieces are made of high quality borosilicate glass
- Set 27 BU/M. Housed in die cut foam lined box
- Kit consists of: 1 Flask (pear shaped, 50 ml), 1 still head, 1 liebig condenser, 1 screwcap adapter, 1 receiver adapter, 1 air leak/steam inlet tube, 1 dropping funnel 50 ml with GP rotaflow tap, 1 thermometer
Stored in die cut foam lined box. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Customers Who Bought This Also Bought