Hoffman Electrolysis Metal Stand
# de catalogue 470344-862
Fournisseur: Wards
Spécifications
- Description:Hoffman electrolysis metal stand
- Cat. no.:470344-862
Spécifications
A propos de cet article
Measuring 24" in overall height, this metal stand with support clamps provides stability and versatility for Hoffman electrolysis experiments.
- 8×5" base is powder-coated
- 23.6" stainless steel rod
Hoffman Electrolysis Metal stand with support clamps is designed to hold components for Hoffman electrolysis equipment.
Customers Who Bought This Also Bought