Adjustable Funnel Stand for 2 Funnels
# de catalogue 470344-464
Fournisseur: Wards
Spécifications
- Description:Adjustable funnel stand for 2 funnels
- Cat. no.:470344-464
Spécifications
A propos de cet article
Designed for two funnels, this polished wood stand features a 1.5" hole diameter and stands 18" tall, providing versatility and stability in the lab.
- Funnel stand featuring a sturdy hardwood construction that will stand up to the wear and tear of the lab
- Height extends up to 18". Base is approximately 11¹/₃ by 4¹/₄"
- Stand comes with clamping screws for adjusting the height
- Designed for use in professional laboratories, but is an ideal choice for science classrooms
Double funnel stand made from quality polished wood. Holds up to two funnels. Hole diameter is 1.5". Height of the funnels is easily adjustable. Features a clamping screw to ensure easy movement when loose and sturdy placement when tightened, and a sturdy wooden base with rubber stoppers for increased stability. Each funnel stand is 12" long, 4.25" wide, and stands 18" tall. Minimum height of funnel attachment is 3" above the base, maximum height is 17" above the base.
Customers Who Bought This Also Bought