A-Frame Retort Base
# de catalogue 470344-084
Fournisseur: Wards
Spécifications
- Description:A-frame retort base
- Cat. no.:470344-084
Spécifications
A propos de cet article
A-frame retort base featuring an 8×8" heavy-duty cast iron base in blue. The base provides stability and support for retort stands and setups in laboratory experiments.
- A-shaped frame retort base made of chemical-resistant cast iron with a blue coating
- The 3-point base weighs 4.4 lbs., providing superb stability for experiments and setups
- Base accommodates 0.5" (14 mm) rods (sold separately) secured with a tightening screw
- Durable and sturdy construction of this set makes it an excellent choice for science classrooms
A-frame retort base lab stands are used in every area of science, especially chemistry, for holding and manipulating an experimental apparatus. This retort stand is made of chemical-resistant cast iron, and features a hole, suitable for rods with a 14 mm metric thread. Stand measures 8" long, 8" wide, and approximately 2" in height. Stand weighs 4.4 lbs., providing superb stability for basic experiments and setups. Sturdy construction and economical cost make this base an excellent choice for science classrooms.
Customers Who Bought This Also Bought