Synthware NMR Tubes, 5 mm
Fournisseur: Kemtech America
Certificats
A propos de cet article
Economy grade NMR tubes with caps included.
- 33 expansion borosilicate glass
- 7"
- 100 MHz rating
Designed to tightly fit in any spinner turbine and have zero NMR background.
Customers Who Bought This Also Bought