Synthware Vacuum or Inert Gas Manifolds
Fournisseur: Kemtech America
Certificats
A propos de cet article
Vacuum manifold with front-left-right and rear-left 10 mm hose connections.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
General purpose double bank manifold for use with vacuum and inert gas.
Customers Who Bought This Also Bought