Synthware High Vacuum Valves, Teflon® Protected, Low Hold-Up
Fournisseur: Kemtech America
Certificats
A propos de cet article
Valves feature two 9 mm O.D. sidearms bent to minimize the hold-up of liquids.
- Heavy wall construction
- 33 expansion Borosilicate glass
Viton® O-Rings on PTFE shaft are protected with precision Teflon® wipers. The plug has a machined-in integral PTFE tip O-Ring to make the primary seal. This design provides an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.
Customers Who Bought This Also Bought