Synthware High Vacuum Valves, PTFE, O-Ring, Three-Way
Fournisseur: Kemtech America
Certificats
A propos de cet article
Valves feature two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another.
- Heavy wall construction
- 33 expansion Borosilicate glass
The non-rotating PTFE plug has three O-Rings. The tip O-Ring makes the primary seal on the precision formed taper seat.
Customers Who Bought This Also Bought