A propos de cet article
VWR® tube rollers are designed for gentle yet complete mixing of a wide range of sample vessels and tubes.
- Adjustable rolling speed
- Accepts tubes and bottles between 8 and 200 mm
- Additional 5° tilting motion for complete mixing
- Timer can be set from 1 minute to 100 hours, with additional continuous operation mode
The VWR® tube rollers are designed for gentle yet complete mixing of a wide range of sample vessels, including Vacutainers™, media bottles, blood tubes, centrifuge tubes and roller bottles. With both models, speed is controlled digitally and is variable between 2 and 80 rpm. In addition to the rolling motion, the VWR® tube rollers also feature a gentle 13 mm (0.5") elevation function on the right-side end of each roller. The tilting feature provides a heightened level of mixing homogeneity and uniformity without placing unnecessary stress on samples. VWR® tube rollers additionally feature an integrated timer, which is controlled digitally. The VWR® tube rollers are the right choice for research laboratories, clinical laboratories and physician's offices in need of a tube mixing solution.