Alnico Bar Magnets
# de catalogue 470349-636
Fournisseur: United Scientific Supplies
Spécifications
- Description:Alnico bar magnet pair
Spécifications
A propos de cet article
Strong Alnico bar magnets for demonstrations.
- Includes a keeper
- Strong Alnico magnets
- Perfect for demonstrating magnetic fields
Powerful Alnico 1 bar magnets, shipped in pairs, with end keepers. Magnets are painted with North and South poles, clearly marked. Each bar is 2" long and ¹/₄" thick.
Customers Who Bought This Also Bought