Equipotential Trough Kit
# de catalogue 470349-678
Fournisseur: United Scientific Supplies
Spécifications
- Description:Equipotential trough kit
- Cat. no.:470349-678
Spécifications
A propos de cet article
Plot equipotential lines accurately with water and a digital multimeter.
- Plot equipotential lines for various electrode configurations
- Classic design
- Correlates to NGSS Standards MS-PS2-5 and HS-PS2-4
- Activity guide included
This comprehensive kit allows students to plot equipotential lines in an electric field between two electrodes held at different electric potentials. A simple setup keeps electrodes partially immersed in water containing an electrolyte, allowing a small current to flow between the electrodes. Students can then sample the resulting electric field using the included multimeter.
Customers Who Bought This Also Bought