United Scientific Aluminum Alloy Tuning Forks
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Long, pure sounding aluminum tuning forks.
- Aluminum alloy
- Each fork is individually stamped with the frequency and note
The frequency and scale letter is stamped on each fork. These easy to activate tuning forks are perfect for investigating sound. Do not strike with a hard object, use a tuning fork activator.
Customers Who Bought This Also Bought