Microwell Plate Label Kits
Fournisseur: Computype
Certificats
A propos de cet article
Durable microplate labels engineered to withstand extreme temperature cycles and chemical exposure.
- Polypropylene labels suitable for automatic application
- TS839P polypropylene, holds up to −80 °C freeze/thaw cycles and an autoclave
- Resistant to solvents such as DMSO, methanol, isopropyl alcohol, and more
The 3'' core is compatible with thermal transfer printers and S3200 Plate Applicator.
Ordering information: Kit includes labels, print ribbon, and cleaning cloth.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.