Sun Track Plastic Hemisphere
# de catalogue 470350-114
Fournisseur: Science First
Spécifications
- Description:Sun track plastic hemisphere, single
Spécifications
A propos de cet article
This device shows how the sun's appearance changes over a period of one month.
Many students believe that the sun is at zenith at noon for every day of the year. By making this model, they will learn the true daily path of the sun. Repeating the same thing one month later, they will compare the daily paths of the sun for different seasons of the year and be active participants in long-duration observations.
Customers Who Bought This Also Bought