Rain Gauge
# de catalogue 470349-728
Fournisseur: United Scientific Supplies
Spécifications
- Description:Rain gauge
- Cat. no.:470349-728
Spécifications
A propos de cet article
Easy to read rain gauge for mounting on a wall or posting in the ground.
- Mounts to a post or staked into the ground
- Measures rainfall up to 5"
- Curved lens for easy reading
Portable rain gauge can be mounted on a wall or staked into the ground. The gauge measures up to 5" of rainfall. Made from durable polystyrene. The large clear increments are easy to read and the curved lens magnifies the look of the numbers and markings when rain is captured. Product dimensions are 12L×2.7W×1.5"D.
Customers Who Bought This Also Bought