Spécifications
- Description:Laboratory starter kit
Spécifications
A propos de cet article
Economical labware starter set.
- 37 total items
- Includes beakers, bottles, cylinders, and flasks
- Borosilicate glass
This convenient set includes an assortment of commonly used glassware for chemistry classrooms and laboratories. The set includes beakers, media bottles, graduated cylinders, and Erlenmeyer flasks, with 37 items in total.
Set includes: (3) 50 ml Beakers, Low Form Griffin, borosilicate; (3) 100 ml Beakers, Low Form Griffin, borosilicate; (6) 250 ml Beakers, Low Form Griffin, borosilicate; (3) 250 ml Erlenmeyer Flasks, Wide Mouth, borosilicate; (6) 500 ml Erlenmeyer Flasks, Wide Mouth, borosilicate; (3) 100 0ml Erlenmeyer Flasks, Wide Mouth, borosilicate; (1) 100 ml Graduated Cylinder, Class B, Double Scale, borosilicate; (1) 250 ml Graduated Cylinder, Class B, Double Scale, borosilicate; ; (1) 500 ml Graduated Cylinder, Class B, Double Scale, borosilicate; (2) 100 ml Media Bottles, GL 45, borosilicate; (3) 250 ml Media Bottles, GL 45, borosilicate; (3) 500 ml Media Bottles, GL 45, borosilicate; (2) 1000 ml Media Bottles, GL 45, borosilicate.