AI Bacteria Analysis Slides
Fournisseur: Apacor
Certificats
A propos de cet article
Fiducial printed glass microscope slides for AI image analysis.
- Printed fiducial design
- White glass
- Cut edges
Slides are available in two formats, with either a circle center or square center.
Customers Who Bought This Also Bought