A propos de cet article
Modern design, complete with incorporated advanced electronics ensures smooth, problem free operation. Touch screen electronics and PLC programmed pulverising cycle maximizes efficiency of operation. Pneumatic lid clamping system for a range of bowls, and an automatic door locking system preventing access to moving elements during the grinding cycle, maximizes safety.
- Reduces samples to a particle size of 90 to 95% passing 75 microns
- Touch screen for more accurate control
- Internal control box for maximum safety
- Power drive is separate to the motor to reduce wear
- Easy to adjust belt tension
- Fully fabricated and welded base and frame for added strength
Alsto's unique design split cabinet makes maintenance and service a simple quick operation with all of the working components accessible within seconds. This enables a maintenance technician to access any of the wear of driving/driven parts directly, reducing down time during scheduled maintenance.
Alsto's PV series has a belt driven universal shaft reducing the vibration and impact stresses on the motor increasing life span of the motor. A full range of spare parts for scheduled maintenance including wear parts, are availabel for maitaining optimum operation of the Pulveriser.
Spécifications
- Catalog No:
- 76470-114
- 76638-276
- 76638-278
- 76638-280
- Model:
- PV2
- PV2
- PV5
- PV5