Alsto JC2013 Jaw Crusher
Fournisseur: KLEN INTERNATIONAL (74) PTY LTD.
Certificats
A propos de cet article
The Alsto JC2013 is a reliable, heavy duty crusher designed for general laboratory applications, ready and able to crush a large variety of sample types quickly and efficiently.
- Ease of installation with the supplied floor fixings
- Large 200×130 mm (8×5'') jaw opening allows for an input
- particle size of up to 110 mm
- Rapid crushing at ~500 kg per hour at usual product size
- Two dust extraction points and efficient drive train guarding to ensure operator safety
- Fast and easy jaw gap adjustment
- Hard wearing, easily replaceable jaw plates and side liners
Spécifications
- Catalog No:
- 76638-290
- 76638-292
- Model:
- JC2013
- JC2013
- Final fineness:
- 6 mm normal product size; 35 mm max. product size
- 6 mm normal product size; 35 mm max. product size
- Material feed size:
- 110 mm (max.)
- 110 mm (max.)
- Throughput:
- 500 kg/h
- 500 kg/h