Alsto DP2000 Disc Pulveriser
Fournisseur: KLEN INTERNATIONAL (74) PTY LTD.
Certificats
A propos de cet article
Simple, robust machine designed for fine grinding of a large variety of sample and ore types. One fixed and one rotating 200 mm grinding plate turning on a horizontal axis, with the sample drawer located at the bottom of the chamber.
- A simple screw adjustment to control product sizing via plate gap setting
- Powerful 4kW, 3 phase drive motor
- Supplied as standard on a base plate suitable for bench/workstation mounting
- Durable chilled cast iron disc plates for long life
The Alsto DP2000 Disc Pulveriser is a simple, robust machine designed for fine grinding of a large variety of sample and ore types. The unit comprises one fixed and one rotating 200 mm grinding plate turning on a horizontal axis. Internal surfaces of the grinding chamber are smooth and easily cleaned, with the sample drawer located at the bottom of the chamber.
Spécifications
- Catalog No:
- 76638-282
- 76638-284
- Model:
- DP2000
- DP2000
- Final fineness:
- 75 micron (min), normal product size
- 75 micron (min), normal product size
- Material feed size:
- 12 mm (max.)
- 12 mm (max.)
- Throughput:
- 15 - 150 kg/h depending on disc gap setting
- 15 - 150 kg/h depending on disc gap setting