Half-Watt Resistors
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Quality resistors for electrical experiments.
- Quality carbon resistors
- Power rating 0.5 W
- Color coded resistance values
- 20 to 25 mm wire leads
These resistors are designed to easily fit into protoboards or breadboards. Each 0.5 W resistor is color coded to designate its electrical resistance. The leads on each end are about 1" long.
Ordering information: These 0.5 W carbon resistors are available in packs of 10 only.
Customers Who Bought This Also Bought