A propos de cet article
The inner doors on these flammable storage freezers protect stored substances with superior cooling retention.
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These flammable storage freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
Our Performance Series includes premium additions such as state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
*Select models ENERGY STAR certified: 76644-708, 76644-712, and 76644-716
Spécifications
- Catalog No:
- 76644-716
- 76644-712
- 76644-708
- Model:
- 76644-716
- 76644-712
- 76644-708
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Defrost:
- Manual
- Manual
- Manual
- Shelves/drawers:
- 7 inner doors
- 7 inner doors
- 7 inner doors
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Weight:
- 119 kg (262 lbs.)
- 113 kg (250 lbs.)
- 91 kg (200 lbs.)
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants