A propos de cet article
The inner doors on these flammable storage freezers protect stored substances with superior cooling retention.
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These flammable storage freezers meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
*Select models ENERGY STAR certified: 76644-706, 76644-710, and 76644-714
Spécifications
- Catalog No:
- 76644-714
- 76644-710
- 76644-706
- Model:
- 76644-714
- 76644-710
- 76644-706
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Defrost:
- Manual
- Manual
- Manual
- Shelves/drawers:
- 7 inner doors
- 7 inner doors
- 7 inner doors
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Weight:
- 118 kg (261 lbs.)
- 113 kg (249 lbs.)
- 90 kg (199 lbs.)
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants