VWR® Plus Manual Defrost Freezers with Inner Doors
Fournisseur: VWR International
Certificats
A propos de cet article
The inner doors on these Plus Series freezers protect stored substances with superior cooling retention.
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
*Select models ENERGY STAR certified: 76644-730, 76644-734, and 76644-738
*Select models ENERGY STAR certified: 76644-730, 76644-734, and 76644-738
Ordering information: All units come with a two-year parts and labor warranty and a five-year compressor parts warranty.
Spécifications
- Catalog No:
- 76644-738
- 76644-734
- 76644-730
- Model:
- 76644-738
- 76644-734
- 76644-730
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Defrost:
- Manual
- Manual
- Manual
- Shelves/drawers:
- 7 inner doors
- 7 inner doors
- 7 inner doors
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Weight:
- 118 kg (261 lbs.)
- 113 kg (249 lbs.)
- 90 kg (199 lbs.)