VWR® Performance Manual Defrost Freezers with Inner Doors
Fournisseur: VWR International
Certificats
A propos de cet article
The inner doors on these Performance Series freezers protect stored substances with superior cooling retention.
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These manual defrost laboratory freezers offer professional cold storage for scientific and industrial settings. The Performance Series showcases our most elite features including state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: ETL Listed
*Select models ENERGY STAR certified: 76644-732, 76644-736, and 76644-740
*Select models ENERGY STAR certified: 76644-732, 76644-736, and 76644-740
Ordering information: All units come with a three-year parts and labor warranty and a five-year compressor parts warranty.
Spécifications
- Catalog No:
- 76644-740
- 76644-736
- 76644-732
- Model:
- 76644-740
- 76644-736
- 76644-732
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Defrost:
- Manual
- Manual
- Manual
- Shelves/drawers:
- 7 inner doors
- 7 inner doors
- 7 inner doors
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Weight:
- 119 kg (262 lbs.)
- 113 kg (250 lbs.)
- 91 kg (200 lbs.)