ChemTrap® H402 Chemical Filtration Option for Safety Cabinets, Erlab
Fournisseur: Erlab
Certificats
A propos de cet article
Add on filtration system that will filter the interior space of a flammable, acid, or corrosive cabinet.
- User protection while saving energy
- Easy installation, connection via bung port - no ductwork needed
- Eliminates toxic fumes and odors while purifying laboratory air
- Adaptable to any cabinet
- Horizontal (H) model
The filters will capture and retain toxic fumes present within the cabinet, and will return clean air back to the laboratory, eliminating odors when the door is opened. Designed as a standalone system with no need for ducting to the outside, it is equipped with an adjustable pipe that accommodates connection points from the side or back of your existing cabinet via the bung. It will come with the needed elbows, sleeves, and threaded port to connect your safety cabinet to the Chemtrap.
Certifications: Meets AFNOR NFX 15-211 filtration efficiency standard and ANSI Z9.5 2012 standard.
Ordering information: All models require a filter to operate. Filters are sold separately. Please contact your Avantor representative for more information.
Spécifications
- Catalog No:
- 76640-766
- 10813-212
- 10812-238
- Airflow rate:
- 35 CFM
- Fans:
- 1
- Electrical:
- Available in 115 V/60 Hz