Mobile Phase Reservoir Bottle, MicroSolv
# de catalogue 97057-216
Fournisseur: MicroSolv Technology Corp
Spécifications
- Capacity:1 L
- Description:Mobile phase reservoir bottle, glass, clear, standard taper top, 29/32 neck
- Cat. no.:97057-216
- Supplier No.:58801-01
Spécifications
A propos de cet article
This borosilicate glass bottle is perfect for HPLC mobile phase reservoirs with its small opening and large capacity. Graduations on the outside of the bottle help you determine the Solvent use and prevent running dry.
This Bottle requires a S/T (Standard Taper) 29/32 joint for the mouth and will produce a tight seal. These bottles are often used on older HPLC Instruments such as the Agilent 1090.
Customers Who Bought This Also Bought