Flux® Heavy-Duty Drum Pump Motors, TEFC
Fournisseur: FLUX PUMPS CORP
Certificats
A propos de cet article
Configurable to your requirements.
- High-viscosity tube set handles fluids with viscosities to 50,000 cp
Heavy-duty motors are best for applications requiring high output head. Motors have a built-in overload switch with manual rest to avoid the danger of automatic restarting.
Order tube set and accessories separately.
Spécifications
- Catalog No:
- MFLX07076-20
- CE regulatory approval:
- Yes
- Max. viscosity:
- 800 cP
- Motor type:
- TEFC
- Motor hp:
- 3/4
- Power:
- 120 VAC; 60 Hz
- Warranty:
- 3 months