VWR® ULT Freezers Eco Premium 3" Cryobox Storage Bundles, 120 V
Fournisseur: VWR International
New Product
Certificats
A propos de cet article
The VWR® ULT Freezers Eco Premium are designed for safe and reliable storage of sensitive samples. They feature ergonomic control and low energy consumption.
- Maximum 3" box capacity: 224/336
- Energy star certified
- Temperatures between −86 to −50 °C
- Long warm-up time (warm up time from −86 to −50 °C: 360 min)
- Highly efficient VIP insulation and long-lasting rust-proof sample chamber
- Green refrigerants
- USB and Ethernet interface for data transfer
The VWR® ULT Freezers Eco Premium have a stainless steel inner chamber, flexible rack configuration and are further characterized by low energy consumption and electricity costs. They have individually adaptable chamber compartments and samples remain frozen for >30 hours during power outage or system failure. The climate-friendly refrigerants used help to protect the environment.
Warranty: VWR® Five-year limited parts and labor warranty; VWR® Five-year warranty on compressor parts.
Certifications: UL, cUL, CE marked, UKCA.
Ordering information: A variety of accessories are available, these must be ordered separately.
Delivery: VWR® ULT Freezers are supplied with racks for 3" standard boxes for full freezer load.
Packaging: VWR® ULT Freezer plus a package of racks for 3" standard boxes.
Spécifications
- Catalog No:
- 76632-850
- 76632-852
- 76632-858
- 76936-168
- 76936-170
- 76632-860
- Environmentally Preferable:
- Low Manufacturing Impact, Energy Efficient
- Low Manufacturing Impact, Energy Efficient
- Low Manufacturing Impact, Energy Efficient
- Low Manufacturing Impact, Energy Efficient
- Cat. No.:
- 76632-850
- 76632-852
- 76632-858
- 76632-860
- Electrical:
- 120 V
- 120 V
- 120 V
- 120 V
- Environmental certification:
- EnergyStar; ISO 14001
- EnergyStar; ISO 14001
- EnergyStar; ISO 14001
- EnergyStar; ISO 14001