A propos de cet article
Boekel Scientific’s large, forced air incubators, with two internal outlets, are customizable for most general incubation needs. Units are equipped with a digital control system with built-in timer and a door with a large viewing window. For light-sensitive incubations, a removable door panel that easily omits ambient light is included.
- Excellent temperature stability, uniformity, and recovery time
- Wide temperature range for multiple applications
- Single door configuration with removable door panel for light and dark incubations
- Bright easy-to-use digital display with built-in timer
- Two internal outlets for multiple accessories
- 2-year warranty
- UL, CE, CSA approved
Boekel Scientific’s line of forced air incubators are ideal for microbiological testing, wastewater and water quality testing, and general incubation needs. The 1.4 and 2.5 cu. ft. sizes (listed separately) are equipped with 1 internal auxiliary outlet, and these 5.0 cu. ft. models feature 2 internal auxiliary outlets. This allows for flexibility and convenience when using additional equipment or accessories in your incubation process. All units come standard with a large viewing window and an easily removable door panel that occludes ambient light. The space saving designs, large temperature range, consistent uniformity, and two-year warranty ensure these incubators will be integral parts of your laboratory for years to come.
Chamber Dimensions (W×D×H) : 43.2×43.2×76.2 cm (17×17×30")
Spécifications
- Catalog No:
- 76644-928
- 76644-918
- Environmentally Preferable:
- Low Manufacturing Impact, Waste Reducing
- Low Manufacturing Impact, Waste Reducing
- Convection type:
- Forced convection
- Forced convection
- Shelves supplied (max.):
- 3 (6)
- 3 (6)
- Environmental certification:
- RoHS/WEEE Complaint
CDP Disclosure - RoHS/WEEE Complaint
CDP Disclosure