Nasco Healthcare® CPR Cube Pro
# de catalogue 470335-352
Fournisseur: Nasco
Spécifications
- Description:CPR cube pro
- Cat. no.:470335-352
Spécifications
A propos de cet article
Compact CPR compression training.
- App feedback for compression rate and depth
- Audible confirmation of correct depth
Wireless connection for both student and instructor App. Supports remote training between student and instructor. App allows for mass instructions without limitations of traditional wireless controls whether on-site or remote.
Customers Who Bought This Also Bought